Purge Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This glass purge adapter is desgined for use with rotary evaporators.
- Top spherical socket joint
- Bottom spherical ball joint
- Middle tube stopcock or valve
- Fabricated from borosilicate glass
Top has a spherical 35/25 socket joint for joining to the condenser or trap, and the bottom has a spherical 35/25 ball joint for joining to the receiver flask. Middle tube has either a 4mm PTFE stopcock, or a 2mm valve to allow for purging of the vapor from the receiver flask or for adding purge gas into the flask. Overall length is 75mm.
Customers Who Bought This Also Bought