Bench Top Personal Workstation, PVC, TrippNT
# de catalogue 11376-294
Fournisseur: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Color:White
- Description:Workstation with 17 Compartments, Holders, and Dispensers
- W×D×H:61×20×30 cm (24×8×12")
- Supplier No.:50014
Spécifications
A propos de cet article
White PVC bench top personal workstation has 17 compartments, holders and dispensers to sort and organize supplies
It fits neatly on the bench or counter and made of bright white chemically resistant PVC construction.
Weight: 2 kg (5 lbs.)
Customers Who Bought This Also Bought