Rotating Multi-Compartment Workstation, PVC, TrippNT
# de catalogue 11376-292
Fournisseur: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Rotating Multi-Compartment Workstation
- W×D×H:36×33×33 cm (14×13×13")
- Weight:5 kg (12 lbs.)
- Supplier No.:50153
Spécifications
A propos de cet article
White PVC rotating multi-compartment workstation sorts, stores and protects a variety of lab supplies
Made from durable white PVC plastic makes it easy to clean. This workstation includes 12 flexible compartments to organize and share supplies or tools. It also has a built in small Kimwipe holder and a four slot tool holder.
Customers Who Bought This Also Bought