Bent Gooseneck Drying Tubes, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Bent gooseneck-type drying tube features a dessicant area that is 140 mm in length.
- Fabricated from borosilicate glass
Ordering information: Stopper is included.
Customers Who Bought This Also Bought