Dispensing Bins, PVC/Acrylic, TrippNT
Supplier: TrippNT
Certificates
About this item
White PVC/acrylic dispensing bins are great for the bench or countertop
Made from chemically resistant PVC and clear acrylic. The removable top allows for easy filling and the hinged clear acrylic front protects and dispenses supplies. Great for pipette tips, swabs, vials, tubes, and other small consumables.
Customers Who Bought This Also Bought