Graduated Cylinder, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These cylinders have a full length, reinforced standard taper 24/40 outer joint at top
Single scale metric graduations. Can be used for distillation receiver.
Ordering information: For stoppers (not included), order 89062-824.
Spécifications
- Catalog No:
- 89054-028
- 89054-030
- 89054-032
- 89054-024
- 89054-026
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall