To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2N
- Clonality:Polyclonal
- Gene ID:7334
- Host:Rabbit
- Isotype:IgG
- Reactivity:Fish
- Antigen symbol:UBE2N
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Ubiquitin-conjugating enzyme E2 N|UBC13|BLU|UbcH-ben|Bendless-like ubiquitin-conjugating enzyme|UbcH13|HEL-S-71
- Storage buffer:Preservative: 0.02% Sodium Azide; Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Mixture of synthetic peptides containing amino acids 2-19 (AGLPRRIIKETQRLLAEP) and 131-148 (TNEAQAIETARAWTRLYA) of Human UBE2N.
- Tested applications:WB
- Purification:Protein G purified
- Cat. no.:89359-804
- Supplier No.:GTX13663
Spécifications
A propos de cet article
Ube2N (ubiquitin-conjugating enzyme E2N) is a novel human ubiquitin-conjugating enzyme with an ability to form a thiol ester linkage with ubiquitin in an ubiquitin-activating enzyme E1-dependent manner. The amino acid sequence showed 80% identity with the Drosophila's bendless gene product (ubiquitin-conjugating enzyme E2). The corresponding transcripts are highly expressed in heart, skeletal muscle, and testis. UBE2N protein may be involved in protein degradation mainly in the muscles and testis.
WB: Use at a concentration of 2 ?g/ml. Detects a band of approximately 17 kDa (predicted molecular weight: 18.24 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: UBE2N
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish