To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Mannose-6-Phosphate Receptor (Cation Dependent)
- Clonality:Polyclonal
- Gene ID:4074
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:MPR46
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:MPR46|MPR 46|MPR-46|SMPR|CD-MPR
- Storage buffer:PBS, no preservatives added
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide: FESTVGQGSDTYIYIFRVCREAGNHTSGAGLVQINKSNGKETVVGRLNETHIFNGS, corresponding to amino acids 60-114 of Human Mannose 6 Phospate Receptor.
- Tested applications:ICC/IF, WB
- Purification:Affinity purified with antigen
- Cat. no.:89367-462
- Supplier No.:GTX16417
Spécifications
A propos de cet article
Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of 300 kDa cation-independent mannose 6-phospate receptors (MPR). MPR cycles and transports newly synthesized lysosomal enzymes between the TGN and late endosomes / early lysosomes. It also cycles and transports extracellular lysosomal enzymes between the plasma membrane and early endosomes via clathrin coated vesicles. The bulk of the MPR protein is in the extracellular/lumenal domain. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPR's is predominantly within late endosomes, fewer in the trans Golgi network and ~5-10 % at the cell surface. In addition to its man6P binding activity, the MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An ~240 kDa soluble, truncated form, representing the extracellular domain of the protein, has also been found circulating in serum and is capable of binding both ligands.
For ICC: Use at a dilution of 1:150 - 1:250. For WB: Use at an assay dependent dilution. Predicted molecular weight: 31 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: MPR46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human