To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 7
- Clonality:Monoclonal
- Clone:7417.111
- Gene ID:3574
- Host:Mouse
- Isotype:IgG1
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:IL7
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:IL-7|IL7|Interluekin-7|Interluekin 7|IL 7|interluekin
- Storage buffer:Preservative: None Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested applications:ELISA, Neut, WB
- Purification:Protein A purified
- Cat. no.:89355-046
- Supplier No.:GTX10765
Spécifications
A propos de cet article
Mouse monoclonal [7417.111] to IL7
ELISA: Use at a concentration of 1 - 2 ?g/ml. Neut: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 ?g/ml. Predicted molecular weight: 17 kDa. The exact concentration of antibody required to neutralize recombinant human IL7 activity is dependent on the cytokine concentration, cell type, growth conditions and the type of activity. The detection limit in immunoblotting for recombinant human IL7 is approximately 25 ng/lane and 300 ng/lane under nonreducing and reducing conditions, respectively. A working concentration of the capture antibody at 1-2 ? g/ml (100?l/well) in combination with a biotinylated polyclonal detection antibody at 25-50 ng/ml (100?l/well) is recommended to detect human interleukin 7. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IL7
Clonality: Monoclonal
Clone: 7417.111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human