To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells Inhibitor, Beta
- Clonality:Polyclonal
- Gene ID:4793
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:NFKBIB
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:50 μl
- Western blot:Yes
- Antigen synonyms:IkBb|ikB-B|IKappaBbeta|I-kappa-B-beta|I(Kappa)B(beta)|IKB-beta|NF-kappa-BIB|IkB
- Storage buffer:0.01% (w/v) Sodium Azide
- Storage temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Immunogen:IkBb peptide corresponding to a region near the C-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Tested applications:ELISA, IP, WB
- Purification:Antiserum
- Cat. no.:89362-606
- Supplier No.:GTX27547
Spécifications
A propos de cet article
Rabbit Polyclonal antibody to IkB beta
For ELISA: Use at an assay dependant dilution. For IP: Use at an assay dependant dilution. For WB: Use at a dilution of 1:1,000. Detects a band of approximately 45 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NFKBIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat