To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Hepatitis C E2
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Reactivity:Virus
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:Phosphate-buffered saline, pH 7.2, containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Recombinant full length HCV E2 protein, genotype 1a
- Tested applications:ELISA, ICC/IF, WB
- Purification:Protein A purified
- Cat. no.:89367-716
- Supplier No.:GTX40852
Spécifications
A propos de cet article
Hepatitis C E2 is a virus envelope glycoprotein which forms a heterodimer with the E1 protein.
Type: Primary
Antigen: Hepatitis C E2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus