To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Clonality:Monoclonal
- Clone:DX2
- Gene ID:355
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:FAS
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
- Storage buffer:Preservative: 0.09% Sodium Azide; Constituents: PBS
- Storage temperature:store at -20 C
- Concentration:2mg/ml
- Immunogen:Mouse L cells transfected with a human Fas/CD95 cDNA.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A purified
- Cat. no.:89350-402
- Supplier No.:GTX10460
Spécifications
A propos de cet article
Mouse Monoclonal antibody [DX2] to Fas
FACS: Use 10?l for 10^6 cells; 1 ug per 100 ul blood. WB: Use at an assay dependent dilution. Predicted molecular weight: 45-48 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone: DX2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human