Laboratory Support, Ace Glass
# de catalogue 89075-728
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Laboratory Support, Poly-Jaque
- Height:3.5 - 12" (adjustable)
- Platform Size:6.5×6.25"
- Weight:15 lbs.
- Supplier No.:11214-20
Spécifications
A propos de cet article
Fabricated from corrosion- and chemical-resistant, glass-reinforced polypropylene and PTFE filled polyethermide, this lightweight support is non-magnetic and non-conducting
It utilizes the scissors jack principle for quick, easy and accurate height adjustment. The PTFE nuts provide smooth, non-freezing operation. The jack can be adjusted in height.
Customers Who Bought This Also Bought