Economy Model Temperature Controller, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Economy Model, compact, digital temperature controller.
16 segment ramp and soak, fuzzy logic, auto tune PID controller. One T, K, or J type sensor input. One 120 V output. Temperature range varies by thermocouple type. Control accuracy of ±0.1 °C. Absolute accuracy of ±0.25 °C of full range. Microprocessor based 1/32 DIN model for digital accuracy and reliability, Bright LED display.
Ordering information: Controller (only) available with "J", "T", or "K" type connections. A complete item, with "J" type connection and Type "J" thermocouple, is also available. Three-year limited warranty. Support stand is not included.
Spécifications
- Catalog No:
- 89076-514
- 71000-056
- 71000-054
- Temperature range:
- –50 to 800 °C, Adjustable
- –50 to 800 °C, Adjustable
- –50 to 800 °C, Adjustable
- Electrical:
- 120V, 50/60Hz, 15A
- 120V, 50/60Hz, 15A
- 120V, 50/60Hz, 15A
- Dimensions:
- 5.25W x 2H x 5.25"D
- 5.25W x 2H x 5.25"D
- 5.25W x 2H x 5.25"D
- Weight:
- 2 lbs., 2 oz.
- 2 lbs., 2 oz.
- 2 lbs., 2 oz.