FoamZorb™ Foam Wipes, Contec
# de catalogue 33500-504
Fournisseur: Contec
Small Business Enterprise
Spécifications
- Dimensions:10 x 13 cm (4 x 5")
- Description:FoamZorb™ Foam Wipes
- Material:Foam
- Cat. no.:33500-504
- Packaging:50 each/bag, 20 bags/case
- Supplier No.:FZ451/8
Spécifications
A propos de cet article
Hydrophilic polyurethane foam, high sorptive capacity with low levels of extractable residues and particles. For spill cleanup or solution/solvent application.
Packaging: 18/bag, 56 Bags/case.
Customers Who Bought This Also Bought