CELLSTAR® Cell-Repellent Surface Microplates and Dishes, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Greiner Bio-One introduces a new surface chemistry designed to effectively inhibit cell adhesion.
- Nonpyrogenic
- Free of RNase, DNase, and human DNA
- Cell-repellent surface
- Ideal substrate for native cell culture experiments
Achieved through an innovative chemical polymer modification, Greiner Bio-One’s cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
The cell-repellent surface is ideal for applications such as spheroid culture of tumor cells, spheroid culture of embryonic cells, embryoid body formation, and suspension culture of semi-adherent and adherent cell lines. All vessels are free of detectable RNase, DNase, and human DNA. nonpyrogenic and noncytotoxic.
Customers Who Bought This Also Bought