High Temperature Graphite Ferrules, Ace Glass
Fournisseur: Avantor
Certificats
A propos de cet article
These single piece, high purity graphite ferrules require no back ferrule, and no organic binders are used
They are ideal for glass-to-glass or glass-to-metal connections. All sizes are used with a #7 Ace-Thred. Due to their high purity and unique properties, these ferrules make a mono-molecular seal — one of the strongest seals known.
Ordering information: Note that ferrules are not always suitable for vacuum applications.
Customers Who Bought This Also Bought