Chromatography Indicator Sprayer, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This convenient, powerful spray unit is used for spraying indicators.
Ordering information: Complete unit consists of spray head with dip tube, can of propellent, and plastic jar. Refill sprays up to one liter.
Customers Who Bought This Also Bought