Three-Way Tubing Connector Valve, Ace Glass Incorporated
# de catalogue 89077-462
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Three -Way Tubing Connector Valve
- Cat. no.:89077-462
- Supplier No.:12730-39
Spécifications
A propos de cet article
This three-way tubing connector valve is used for applications where stream switching, reagent switching, sample removal, or sample injection is desirable
The body is machined from a single piece of PTFE. The Tefzel valves are rated to 500psi, and they have one moving part per channel to ensure trouble-free operation and a long service life. The 1.5mm I.D. channels meet at valve center. Leak-tight to outside even at excessive pressures. Dead space within the valve is zero. Caps are polypropylene with Fluoron elastomer O-Ring seals for instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D. Equipped with ¼"–28 UNF thread at the back of the caps for direct connection with other adapters. This valve is autoclavable.
Customers Who Bought This Also Bought