Two-Way Tubing Connector Valve, Ace Glass Incorporated
# de catalogue 89097-278
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Two-Way Tubing Connector Valve
- Cat. no.:89097-278
- Supplier No.:12728-32
Spécifications
A propos de cet article
Similar to the Variable Bore Tubing Connector (89077-438), except this Two-Way Tubing Connector Valve features a single Tefzel control valve rated at 500psi
The valve has one moving part for trouble-free operation and a long service life. It's unique design prevents the valve from leaking, even at excessive pressures. The body is machined from a single piece of PTFE. Caps are polypropylene with Fluoron elastomer seals for instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D. Equipped with ¼"–28 UNF thread at the back of the cap for direct connection to other adapters. This valve is autoclavable.
Customers Who Bought This Also Bought