Miniature Valve, Inert, Ace Glass Incorporated
# de catalogue 89052-924
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Miniature Valve, Inert
- Cat. no.:89052-924
- Supplier No.:5850-10
Spécifications
A propos de cet article
This miniature straight-through shut-off valve is designed for small volumes of ultrapure or corrosive fluids
Liquids touch only inert PTFE or inert CTFE. The housing features ¼"–28 UNF female threaded ports with 0.059" I.D. holes to accept a variety of fittings.
Customers Who Bought This Also Bought