FETFE O-Ring Replacement Packs for Stopcocks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These FETFE O-Ring replacement kits are designed for use with stopcock plugs 89062-434, 89062-436, and 89062-490 (among others)
For Kalrez equivalents, see items 89060-900 (0–3), 89060-906 (0–5, 0–8), or 89060-918 (0–10).
Customers Who Bought This Also Bought