A propos de cet article
The Roxy M™ Repeating Pipettor provides precise, accurate, and reliable repetitive pipetting of volumes from 1 μl to 5 ml.
- Lightweight, ambidextrous design
- Comfortable grip
- Virtually maintenance-free with no internal wear-and-tear parts
- Robust construction
Its ergonomic design is slim, lightweight (105 g), and features an ambidextrous design for a comfortable grip.
Factory calibrated with no internal wear-and-tear parts, the fully manual Scienceware® Roxy M™ Repeating Pipettor is maintenance-free. Each pipettor is identified with a serial number and comes with an adapter for use with 25 and 50 ml tips (adapter pack 89233-700 also available separately).
Use with Scienceware® Roxy M™ Tips (available in nonsterile and sterile formats covering a wide range of sizes) to offer nearly 60 different volume settings in as many as 48 steps without refilling the syringe tip (depending on the tip size and pipettor volume) all while providing accurate, precise, and reliable pipetting every time.
A well-balanced feel and convenient layout provides one-handed volume selection and smooth dispensing. The robust construction and positive displacement operation results in accurate and precise dispensing of a wide range of liquids, including high viscosity or high vapor pressure liquids. Select the desired volume setting by turning the adjustment wheel, fill the tip by sliding the fill lever, and repeatedly dispense the desired volume in intervals as fast as one per second.