BRAND BRANDplates® pureGrade™ Microplates, Nonsterile, BrandTech®
Fournisseur: Brandtech
Certificats
A propos de cet article
BRANDplates® pureGrade™ plates are nonsterile, untreated plates designed for a variety of standardized applications
- Untreated, "medium binding" plates
- ANSI-SLAS formatted
- Non-sterile
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. White and black 96-Well plates feature a gray embossed alphanumeric code for easy well identification.
Made of virgin polystyrene (89093-728 made of proprietary UV-transparent polymer), these plates are suitable for homogenous assays, screening, and storage.
Ordering information: The 96-Well plates are supplied in packs of 100. The 384- and 1536-Well plates are supplied in packs of 50.
Customers Who Bought This Also Bought