![fluid-handling-US&CA-hero1920x760.jpg](https://occapi.avantorsciences.com/medias/fluid-handling-US-CA-hero1920x760.jpg?context=bWFzdGVyfHJvb3R8MTg5NDEzfGltYWdlL2pwZWd8YUdaaUwyaGhaaTg1TkRjNU56ZzNNekUxTWpNd0wyWnNkV2xrTFdoaGJtUnNhVzVuTFZWVEprTkJMV2hsY204eE9USXdlRGMyTUM1cWNHY3wxMmIwNmI5MmQ4NzBlODA5OThmOTU5OWVkOTc2YTIxNTk3YzQ5MjkzOTAzYjRlZTZkMTcxMmI3MmUwNmQ3NTM0)
Fluid Handling
Avantor® is the only open-architecture fluid handling provider to offer complete design, manufacturing and logistics to support every stage of your biomanufacturing process – wherever your operations are located across the globe.
An extensive portfolio of pump technologies to efficiently transfer or dispense all types of fluids.
Shop for a variety of pump and transfer tubing, fittings, gaskets, sanitary clamps and more.
Complete control of your flow path with valves, flow indicators and switches, fluid sensors, flow meters and more.
Fluid handling assemblies and systems to eliminate risk of cross-contamination and the need for time-consuming sterilization in your bioprocessing workflows.
![Peristaltic-Pump-Fluid-Handling955x590.jpg](https://occapi.avantorsciences.com/medias/Peristaltic-Pump-Fluid-Handling955x590.jpg?context=bWFzdGVyfHJvb3R8NDkxNjZ8aW1hZ2UvanBlZ3xhRFpqTDJobE1pODVORGM1Tnpnek9UUXdNVEkyTDFCbGNtbHpkR0ZzZEdsakxWQjFiWEF0Um14MWFXUXRTR0Z1Wkd4cGJtYzVOVFY0TlRrd0xtcHdad3wzNjRjOTYwOTRlYjMwZTAzNDU1ZTcyZWYwNWQ3MzczN2UxODYyNmMyMzkwNjg0ODI2OWRhNDE0NzQxYzE4NDY2)
Masterflex® Peristaltic Pump and Fluid Handling Solutions
Masterflex® peristaltic pump systems are engineered for a long life. They provide superior accuracy and repeatability, ensuring optimal performance from the research phase to scaled-up production.
Masterflex® I/P® MasterSense™ Process Drives with integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow.
Optimize your processes and reduce risk of contamination with our wide variety of fluidics and single-use products. From media prep to fill finish.
The webinar demonstrates a 4-step process, to develop a highly efficient and accurate sampling system.
Work directly with our pump experts to develop the exact OEM pump and tailored components to meet your exact needs. At Masterflex, we are fluid transfer specialists, and we’re here to ensure your total satisfaction.
Masterflex® peristaltic pumps are a top choice for low-shear pumping. Masterflex® CytoflowTM peristaltic pump heads feature a unique convex roller and concave tube bed that reduces shear and has been shown to improve yields while minimizing damage to the viral contents of a vaccine. Plus easy-to-clean stainless steel housing options.
![Global-fluid-handling955x590.jpg](https://occapi.avantorsciences.com/medias/Global-fluid-handling955x590.jpg?context=bWFzdGVyfGltYWdlc3wxMTEwNzN8aW1hZ2UvanBlZ3xhR016TDJoa05TODVOVEl3T1RjeE56UXpNall5TDBkc2IySmhiQzFtYkhWcFpDMW9ZVzVrYkdsdVp6azFOWGcxT1RBdWFuQm58M2YxZDJkMzk2ZWU2OGE4NjRlZWQ3ZTc5NzkzNWIxZjU0Y2I1ZTE5ZWI5ODcxNGFlMTQ3MGQ1N2E3MjNiZTMwYg)
Avantor Fluid Handling: Experts in end-to-end fluid management solutions
Components engineered for precision, compatibility, compliance, and traceability. Large selection to fit your requirements and scale from lab to production.
Masterflex® MasterSense™ pumps with intuitive interface, integrated pressure sensor, and more advanced features for today's demanding processes.
Single-use custom assemblies offer complete design, manufacturing and logistics solutions, ISO class 7 certified cleanroom.
Broad selection of fluid path tubing formulations and sizes, fittings, sensors, controllers, and more. Including non-invasive and single-use options.