![XRF1920-760.jpg](https://occapi.avantorsciences.com/medias/XRF1920-760.jpg?context=bWFzdGVyfHJvb3R8Mzg5MjV8aW1hZ2UvanBlZ3xhRGhoTDJoaU5DODVORGM1TnpZMk56TTJPVEkyTDFoU1JqRTVNakF0TnpZd0xtcHdad3xkMTdlN2Y5ZDE0YzVmZmM5ZDU4OTA2MGQ3MGU3NzdlMjQ1N2RmYWEwZGE4NmFlNTk2Y2Q3YjAyNTI2ZTc1ZDQ3)
X-Ray Fluorescence Spectroscopy (XRF)
X-ray fluorescence spectroscopy, or XRF, lets analysts obtain qualitative and quantitative information by measuring the fluorescent X-rays that are emitted from a sample when X-rays are projected onto the sample from an outside source. Avantor® can provide you with the products and materials you need to conduct XRF in your lab. Choose the products that work best for you from our extensive selection.
Spectromelt® fluxes for fusion sample preparation. Lithium borates, Spectromelt A, sodium borates and Spectromelt B10 are available.