ContactUsLinkComponent
HPLC​ | Avantor
HPLC
The right choice of chemical is crucial for your chromatography process. Avantor® offers an extensive range of chemicals to meet your chromatography needs.
What can I do to improve separations in LC-MS?
You can use additives such as trifluoroacetic acid or formic acid to improve the resolution. A complete range of high purity LC-MS additives is available in bottles or ampoules.
What is the role of ion pair reagents in HPLC?
Ion pair reagents rely upon the addition of ionic compounds to the mobile phase to promote the formation of ion pairs with charged analytes. These reagents are comprised of an alkyl chain with an ionizable terminus. When used with common hydrophobic HPLC phases in the reversed phase mode, ion pair reagents can be used to selectively increase the retention of charged analytes.
What are the main causes of ghost peaks in gradient HPLC?
  • Transition of the sample from an auto-sampler to an instrument, especially when impurities are absorbed to the inside and outside surfaces of the needle dipped into the vial
  • If an impurity in the sample has the same absorption wavelength as the compound of interest
  • The water used in the mobile phase is not pure or fresh enough
  • Old mobile phase
  • Poor quality buffers or additives added in the mobile phase
  • Phthalates or plastic additives contained in the plastic containers used during mobile phase preparation
What are the best recommendations for avoiding ghost peaks?
  • Try to use pre-mixed solvents
  • Use fresh eluants with salts
  • Filter your HPLC solvents
  • Use high quality water (18.2 MOhm, TOC<5 ppb) or gradient grade water
  • Use good quality buffers and salts
  • Control glassware cleaning
Why is it important to control the pH in the mobile phase?
When samples contain ionizable compounds, mobile phase pH can be one of the most variable in the control of retention in reversed phase RP HPLC separation. However, if it is not controlled properly, pH can be a source of many problems. Since most compounds analyzed by RP HPLC contain one or more acidic or basic functional groups, most mobile phases require pH control. For this reason, buffers are widely used.
Avoid “ghost peaks” in reversed-phase HPLC mode gradient with our large selection of HPLC solvents for laboratory applications.
A full catalog of LC-MS solvents for laboratory applications where a low level of ionic background and low alkaline metal content are essential.
Shop for high-quality LC-MS additives and buffers to improve separation.
See our extensive offering of LS-MS mixes for the pre-blended chemicals you need to minimize baseline noise.
High-quality HPLC ion pair reagents with transparency down to 200 nm for the lab.
All the solvents you need for preparative chromatography. Extremely low evaporation residue, low water content and high purity.
Additional Resources
Explore a full range of chromatography solutions and application notes on our chromatography site.