Anti-CAVIN1 Rabbit Recombinant Polyclonal Antibody (FITC)
Catalog # 77796-186
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Caveolae-associated protein 1 protein (12-44AA)
- Antigen symbol:CAVIN1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q6NZI2
- Antigen synonyms:2310075E07Rik|AW546441|Cavin 1|Cavin|Cavin-1|CAVIN1|CGL4|FKSG13|FLJ90031|MGC118550|OTTMUSP00000002049|Polymerase I and transcript release factor|Ptrf|PTRF_HUMAN|RNA polymerase I and transcript release factor|RP23-279L23.6|TTF I interacting peptide 12
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CAVIN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together