REACTIVE YELLOW 3 IMMOBILIZED 5ML
Catalog #: CA77448-824
Supplier: G-Biosciences
New Product
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36