L ASPARAGINE COLUMN IMMOBILIZED 5ML
Catalog #: CA77448-294
Supplier: G-Biosciences
New Product
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
American White Cross First Aid® Planets and Stars Bandages, DUKAL™ Corporation
Supplier: Dukal
$12.89 to $154.51
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$435.47 to $1,186.77
Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$218.47 to $218.47