TRANSMITTER 0-10PSI
Catalog #: MFLX19406-64
Supplier: PENDOTECH
New Product
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$78.90 to $874.91
Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis), VWR® ACS, Reagent Grade for histology
Supplier: VWR International
$250.27 to $860.06
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$233.35 to $2,174.33
Xylene (mixture of isomers), low water content 98.5-99.41% (isomers plus ethylbenzene) ACS, VWR Chemicals BDH®
Supplier: VWR International
$444.83 to $1,603.99
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$66.72 to $75.62