VIAL FILTR <30% .45UMNYL PNK 450UL CS100
Catalog #: 77391-846
Supplier: THOMSON INSTRUMENT CO
New Product
VIAL FILTR <30% .45UMNYL PNK 450UL CS100
Catalog #: 77391-846
Supplier: THOMSON INSTRUMENT CO
New Product
Customers Who Bought This Also Bought
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
27,11 $ to 19 511,36 $
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Supplier: Air-Tite
33,41 $ to 131,03 $
Conductivity Standards, Potassium Chloride (KCl) Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
222,01 $
Sodium acetate, anhydrous ≥99.0% (by perchloric acid titration), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
764,06 $