Stainless Steel Raised Wire Floors, Tecniplast®
Catalog #: 77322-358
Supplier: Tecniplast Usa
New Product
Specifications
- Description:Raised floor for cage size E, stainless steel wire
Specifications
About this item
Stainless steel design with 30 mm elevation separates animals from bedding and waste.
Not required for standard operation but recommended for post-surgical animals or for those on restrictive diets. Help prevent foot pad swelling. Convenient handle design for easy insertion and removal. Autoclavable.
Customers Who Bought This Also Bought