Reusable Filter Animal Cage Covers, Tecniplast®
Catalog #: 77322-412
Supplier: Tecniplast Usa
New Product
Specifications
- Description:Covers for cage size P
Specifications
About this item
Securely fit Tecniplast® plastic animal cages.
- Filter covers permit atmospheric gas passage with minimal buildup of temperature and humidity
- Covers are constructed of transparent polysulfone with a stainless steel filter retainer frame
- Covers fit over water bottle
Replacement filters are ready-to-use and presized to fit neatly inside the cover.
Customers Who Bought This Also Bought