PROCLEAR PP DEMI B 3UM PEPPB03HHPS3 PK3
Catalog #: 77296-394
Supplier: PARKER HANNIFIN TE
New Product
PROCLEAR PP DEMI B 3UM PEPPB03HHPS3 PK3
Catalog #: 77296-394
Supplier: PARKER HANNIFIN TE
New Product
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
$1,747.88
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$121.35 to $993.13
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$464.49