Erlenmeyer Flasks with Joint
Catalog #: 470353-634
Supplier: United Scientific Supplies
New Product
Specifications
- Capacity:125 ml
- ST Joint No.:24/40
- Cat. no.:470353-634
Specifications
About this item
Erlenmeyer flasks that can use a glass stopper.
- Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
- Flasks include ground joints manufactured per ASTM E-1404, Type 2, Class-1 requirement
- Flasks are printed in white color enamel
Borosilicate glass flasks are designed to provide the best combination of thermal shock resistance and physical strength. They feature 24/40 jointed sockets and graduations in durable white enamel.
Customers Who Bought This Also Bought