Disposable Borosilicate Glass 7.0 Pasteur Pipets
Catalog #: 470353-284
Supplier: United Scientific Supplies
New Product
Specifications
- Capacity:2 ml
- Sterility:Non sterile
- Description:Disposable Pasteur pipets, with cotton plug
- Material:Borosilicate glass 7.0
- Overall Length:229 mm
Specifications
About this item
Disposable Pasteur pipettes are ideally suited for use in both clinical and industrial laboratories.
- Manufactured from Borosilicate glass 7.0
- One-time use eliminates the danger of cross-contamination
- 2 ml capacity
- Available in plain or cotton plugged options
Disposable Pasteur pipettes provide an efficient means for inoculating culture media and transferring samples and reagents in both clinical and industrial laboratories. These 2 ml capacity, non sterile, and disposable pipettes are manufactured from borosilicate glass 7.0. Available in two sizes, with or without cotton plugs.
Customers Who Bought This Also Bought