VWR® Cleanroom Comfort Headbands, Made in USA
Catalog #: 77257-036
Supplier: VWR International
New Product
Specifications
- Clothing Size:M
- Garment application:Cleanroom
- Description:VWR® cleanroom comfort headband
- Fabric:Continuous filament fiber stretch nylon
Specifications
About this item
Sweat absorbing, user-friendly, disposable, and hygienic solution.
- Premium soft comfort fit
- Continuous filament fiber stretch nylon
- Sweat absorbing
- Cleanroom compatible
- Disposable
- Made in USA
The cleanroom comfort headband is designed for comfort and intended for use under a bouffant hat in cleanroom environments. These headbands are tailor made to ensure user comfort, crafted from continuous filament fiber, stretch nylon for a soft and comfortable cleanroom garment. This product minimizes the risk of contamination and eliminates the need for employees to use external products during business operations.
Customers Who Bought This Also Bought