POWER CABLE USB WITH FERRITE
Catalog #: 76924-154
Supplier: Orflo
New Product
Customers Who Bought This Also Bought
Phenol ≥99% saturated solution in water, Biotechnology Grade, buffered
Supplier: VWR
$363.73 to $1,729.17
Kimwipes Push-Up Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$151.21 to $172.13
Chloroform-[D₁] (99.8% D) (Isotopic) +1% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
$124.00
Xylene (mixture of isomers), low water content 98.5-99.41% (isomers plus ethylbenzene) ACS, VWR Chemicals BDH®
Supplier: VWR International
$444.83 to $1,603.99