VWR® Max Protection High ESD Cleanroom Lab Coats
Catalog #: 76919-400
Supplier: VWR International
New Product
Specifications
- Clothing Size:3XL
- Risk area:ESD
- Color:White
Specifications
About this item
Cleanroom ESD lab coats are available with either a front snap closure or a front zipper closure.
- Lab coat featuring high ESD properties
- Lightweight comfort fit
- Snap or zipper closure options available
VWR® Max Protection high ESD cleanroom lab coats feature a collar, three pockets, elastic wrists, and serged seams. Garments are made from 63 gSF, and are packaged 25 per case.
Customers Who Bought This Also Bought