To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:Mouse IgG H&L
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:1 mg
- Western blot:Yes
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Packaging:Plastic vial
Specifications
About this item
Sheep anti-mouse IgG H&L secondary antibody for WB, IHC, ICC and ELISA with samples derived from Mouse.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: WB: 1:5,000 to 1:50,000, IHC: 1:200 to 1:5,000, ICC: 1:200 to 1:5,000, ELISA: 1:10,000 to 1:100,000
Type: Secondary
Antigen: Mouse IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse