nano|Filter Vials, Thomson Instrument Company
Catalog #: 76808-590
Supplier: THOMSON INSTRUMENT CO
Specifications
- Pore size:0.2 µm
- Color:Gray
- Membrane:PES
Specifications
About this item
Thomson nano|Filter Vials offer a low dead volume, allowing filtration with as little as 10 µl of sample with enough remaining filtrate to make a 2 µl injection.
- In vial filtration with minimal sample loss, 10 µl sample for a 2 µl injection
- Minimize sample loss by eliminating multiple transfers
- Compatible with standard UHPLC or HPLC autosamplers (12×32 mm)
- Available in 0.2 µm and 0.45 µm pore sizes
- Split and non-split septums available
Minimal sample loss makes the nano|Filter Vials ideal for critical samples replacing syringe filters, spin columns, autosampler vial, low volume insert, and cap.
Simply add the sample and sample buffer, press, and it is ready for the UPLC or HPLC.
BodyTextAccessories: The accessory Filter Vial Presses quickly depress autosampler-ready filter vials.
Customers Who Bought This Also Bought