SCREWCAP LT BL PTFE/SIL.SEPT 13MM CS1000
Catalog #: 76808-522
Supplier: THOMSON INSTRUMENT CO
New Product
SCREWCAP LT BL PTFE/SIL.SEPT 13MM CS1000
Catalog #: 76808-522
Supplier: THOMSON INSTRUMENT CO
New Product
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$105.11 to $2,771.71
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Cytology Funnels for Shandon CytoSpin™ Centrifuge, Biomedical Polymers
Supplier: Biomedical Polymers
$312.93 to $2,768.11
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
$53.39 to $257.21
Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Supplier: Thermo Scientific Chemicals
$106.00
Borosilicate Capillary Micro Glass Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$121.36 to $137.89