CAP CRIMP ALUMINUM 8MM S/T SOFT PK100
Catalog #: TS6PCC8ST101
Supplier: Thermo Scientific
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$233.35 to $2,174.33
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
$607.92 to $2,080.63
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$78.90 to $874.91