Order Entry
ContactUsLinkComponent
STANDARD SILICONE VISCOSITY RT10 500ML
Viscosity Standards, Silicone Oils, Cannon Instruments
Catalog #: CA10021-858
Supplier:  Cannon
CAS Number:  
Viscosity Standards, Silicone Oils, Cannon Instruments
Catalog #: CA10021-858
Supplier:  Cannon
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Viscosity standard RT10
  • Size:
    500 ml
  • Viscosity:
    (25°C) 9.4 mPa·s

Specifications

About this item

CANNON silicone standards are intended for calibration of brookfield type rotational viscometers.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Certifications: Traceable to NIST.
Customers Who Bought This Also Bought