General Purpose Viscosity Standards, Cannon Instruments
Catalog #: CA10021-224
Supplier: Cannon
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Viscosity standard S200
- Size:500 ml
- Viscosity:(20°C) 460 mPa·s
Specifications
About this item
These certified standards are useful for calibration and verification procedures.
- It is suitable for all types of viscometers
- It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Customers Who Bought This Also Bought