CAP SCREW 9MM GR PP T/S/T PK100
Catalog #: TS6PSC9TSTG
Supplier: Thermo Scientific
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
194,14 $ to 694,12 $
Brilliant II SYBR® MM with ROX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
615,00 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
43,20 $ to 44,79 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Supplier: Biotix
81,72 $ to 326,77 $