CAP SCREW YLW PP 9MM RR/T PK100
Catalog #: TS6ASC9RT1Y
Supplier: Thermo Scientific
Customers Who Bought This Also Bought
Sodium thiosulfate 0.10 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$153.23 to $602.78
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Extran® 300 Liquid Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$363.65 to $1,161.45
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
$213.55 to $2,164.21