WHEATON® Serum Vials
Catalog #: 76716-338
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap included:No
- Capacity:3 ml
- Color:Amber
- Description:Serum vial
- Material:Glass
- Cat. no.:76716-338
- Diameter ×H:16.75×37.7 mm
Specifications
About this item
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Customers Who Bought This Also Bought