ContactUsLinkComponent
FINNTIP 10ML PK 100
Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 76670-568
Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 76670-568
Specifications
  • Volume:
    1000 - 10000 µL
  • Packed:
    100/Bag
  • Cat. no.:
    76670-568
  • Tip Style:
    Finntip™ 10 mL
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought